Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC100131000
- Entrez Gene ID
-
100131000
- Gene Name
- uncharacterized LOC100131000
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK124361.1 |
- |
1 - 1 |
1743 |
63 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU784007.1 |
- |
1 |
|
|
Chr. 17 - 78844716 - 78845289 [-] on Build GRCh38 |
63 |
GenBankmRNA |
AI683153.1 |
- |
1 |
|
|
Chr. 17 - 78844899 - 78845279 [-] on Build GRCh38 |
63 |
GenBankmRNA |
R89497.1 |
- |
|
238 |
|
Chr. - - [-] on Build |
63 |
GenBankmRNA |
HY359061.1 |
- |
1 |
|
|
Chr. 17 - 78844749 - 78845271 [-] on Build GRCh38 |
63 |
GenBankmRNA |
BX093333.1 |
- |
1 |
240 |
|
Chr. 17 - 78844703 - 78845286 [+] on Build GRCh38 |
63 |
GenBankmRNA |
DB351930.1 |
- |
1 |
|
|
Chr. 17 - 78844731 - 78845271 [-] on Build GRCh38 |
63 |
GenBankmRNA |
BM557695.1 |
- |
1 |
454 |
|
Chr. 17 - 78844489 - 78845287 [+] on Build GRCh38 |
63 |
GenBankmRNA |
AI198012.1 |
- |
1 |
|
|
Chr. 17 - 78844848 - 78845271 [-] on Build GRCh38 |
63 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ubiquitin specific peptidase 36 |
USP36 |
DUB1 |
-2.3kb
|
Anti-sense |
Chr. - 78787381 - 78840898 on Build GRCh38 |
Downstream |
TIMP metallopeptidase inhibitor 2 |
TIMP2 |
CSC-21K, DDC8 |
7.7kb
|
Anti-sense |
Chr. - 78852977 - 78925390 on Build GRCh38 |
View Full Product Details