Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100132850
Entrez Gene ID
100132850
Gene Name
uncharacterized LOC100132850
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK126791.1 - 1 - 1 3052 74


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream family with sequence similarity 198 member A FAM198A C3orf41 -13.5kb Sense Chr. - 42979267 - 43057715 on Build GRCh38
Downstream protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-) POMGNT2 AGO61, C3orf39, GTDC2, MDDGA8 4.1kb Anti-sense Chr. - 43079229 - 43106092 on Build GRCh38

View Full Product Details