Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- VN2R9P
- Entrez Gene ID
-
648315
- Gene Name
- vomeronasal 2 receptor 9 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 58040768 - 58066959 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC067882.1 |
- |
1 - 2 |
218 |
91 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CB990186.1 |
- |
1 - 2 |
245 |
|
Chr. 11 - 58047255 - 58060141 [-] on Build GRCh38 |
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
olfactory receptor family 9 subfamily Q member 1 |
OR9Q1 |
|
|
Anti-sense |
Chr. - 58023881 - 58181587 on Build GRCh38 |
Downstream |
olfactory receptor family 9 subfamily I member 1 |
OR9I1 |
OR11-228 |
58.4kb
|
Sense |
Chr. - 58118500 - 58119444 on Build GRCh38 |
View Full Product Details