Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AVP
Entrez Gene ID
551
Gene Name
arginine vasopressin
Gene Aliases
ADH, ARVP, AVP-NPII, AVRP, VP
Chromosome Location
Chr.20: 3082555 - 3093521 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011529267.1 XP_011527569.1 2 - 2 1195 74
GenBank mRNA AF031475.1 - 1 - 1 353 74


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream oxytocin/neurophysin I prepropeptide OXT OT, OT-NPI, OXT-NPI -12.0kb Anti-sense Chr. - 3068871 - 3072517 on Build GRCh38
Downstream UBOX5 antisense RNA 1 UBOX5-AS1 21.5kb Anti-sense Chr. - 3106911 - 3150867 on Build GRCh38

View Full Product Details