Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GHR
Entrez Gene ID
2690
Gene Name
growth hormone receptor
Gene Aliases
GHBP, GHIP
Chromosome Location
Chr.5: 42423775 - 42721878 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242406.2 NP_001229335.1 1 - 1 142 92
GenBank mRNA Z11850.1 - 1 - 1 144 92


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC101926960 LOC101926960 -390.1kb Anti-sense Chr. - 42155833 - 42175345 on Build GRCh38
Downstream coiled-coil domain containing 152 CCDC152 CH5400 190.9kb Sense Chr. - 42756818 - 42802437 on Build GRCh38

View Full Product Details