Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LPCAT4
- Entrez Gene ID
-
254531
- Gene Name
- lysophosphatidylcholine acyltransferase 4
- Gene Aliases
-
AGPAT7, AYTL3, LPAAT-eta, LPEAT2
- Chromosome Location
-
Chr.15: 34358888 - 34367196 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK001119.1 |
- |
1 - 1 |
946 |
94 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB343074.1 |
- |
1 |
|
|
Chr. 15 - 34364607 - 34365191 [+] on Build GRCh38 |
94 |
GenBankmRNA |
BF801913.1 |
- |
1 |
310 |
|
Chr. 15 - 34364871 - 34365302 [-] on Build GRCh38 |
94 |
GenBankmRNA |
AU146575.1 |
- |
1 |
|
|
Chr. 15 - 34364607 - 34365151 [+] on Build GRCh38 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
NUT midline carcinoma family member 1 |
NUTM1 |
C15orf55, FAM22H, NUT |
-6.9kb
|
Anti-sense |
Chr. - 34343315 - 34357737 on Build GRCh38 |
Downstream |
golgin A8 family member A |
GOLGA8A |
GM88 |
13.1kb
|
Sense |
Chr. - 34379068 - 34437466 on Build GRCh38 |
View Full Product Details