Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BMP2K
- Entrez Gene ID
-
55589
- Gene Name
- BMP2 inducible kinase
- Gene Aliases
-
BIKE, HRIHFB2017
- Chromosome Location
-
Chr.4: 78776378 - 78916365 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK021620.1 |
- |
1 - 1 |
1252 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 5096 |
MIR5096 |
|
-66.4kb
|
Sense |
Chr. - 78820752 - 78820821 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1088 |
LINC01088 |
|
82.5kb
|
Sense |
Chr. - 78971748 - 79308799 on Build GRCh38 |
View Full Product Details