Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TCF12
- Entrez Gene ID
-
6938
- Gene Name
- transcription factor 12
- Gene Aliases
-
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
- Chromosome Location
-
Chr.15: 56918090 - 57291261 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK022018.1 |
- |
1 - 1 |
1254 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC145783 |
LOC145783 |
|
-153.7kb
|
Anti-sense |
Chr. - 56886170 - 56918499 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 926 |
LINC00926 |
|
226.0kb
|
Sense |
Chr. - 57300365 - 57307769 on Build GRCh38 |
View Full Product Details