Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KRR1
- Entrez Gene ID
-
11103
- Gene Name
- KRR1, small subunit processome component homolog
- Gene Aliases
-
HRB2, RIP-1
- Chromosome Location
-
Chr.12: 75497639 - 75511638 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK022133.1 |
- |
1 - 1 |
948 |
69 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX101828.1 |
- |
1 |
449 |
|
Chr. 12 - 75510338 - 75511104 [-] on Build GRCh38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
GLI pathogenesis related 1 |
GLIPR1 |
CRISP7, GLIPR, RTVP1 |
-7.7kb
|
Anti-sense |
Chr. - 75480733 - 75501936 on Build GRCh38 |
Downstream |
pleckstrin homology like domain family A member 1 |
PHLDA1 |
DT1P1B11, PHRIP, TDAG51 |
513.8kb
|
Sense |
Chr. - 76025447 - 76031776 on Build GRCh38 |
View Full Product Details