Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RALGAPA1
Entrez Gene ID
253959
Gene Name
Ral GTPase activating protein catalytic alpha subunit 1
Gene Aliases
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
Chromosome Location
Chr.14: 35538352 - 35809304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283043.1 NP_001269972.1 8 - 9 1194 80
NM_001283044.1 NP_001269973.1 8 - 9 1194 80
NM_014990.1 NP_055805.1 8 - 9 1194 80
NM_194301.2 NP_919277.2 8 - 9 1194 80
XM_005267491.3 XP_005267548.1 8 - 9 1272 80
XM_005267492.3 XP_005267549.1 8 - 9 1272 80
XM_005267493.3 XP_005267550.1 8 - 9 1272 80
XM_006720098.2 XP_006720161.1 8 - 9 1272 80
XM_006720099.2 XP_006720162.1 8 - 9 1272 80
XM_006720100.3 XP_006720163.1 8 - 9 1272 80
XM_006720101.2 XP_006720164.1 8 - 9 1272 80
XM_006720102.3 XP_006720165.1 8 - 9 978 80
XM_006720103.2 XP_006720166.1 8 - 9 1272 80
XM_006720104.3 XP_006720167.1 8 - 9 1272 80
XM_011536615.2 XP_011534917.1 8 - 9 1272 80
XM_011536616.2 XP_011534918.1 8 - 9 1272 80
XM_017021141.1 XP_016876630.1 8 - 9 1272 80
XM_017021142.1 XP_016876631.1 8 - 9 1272 80
XM_017021143.1 XP_016876632.1 8 - 9 1272 80
XM_017021144.1 XP_016876633.1 8 - 9 1272 80
XM_017021145.1 XP_016876634.1 8 - 9 1272 80
GenBank mRNA AB020691.1 - 3 - 4 322 80
AB511280.1 - 8 - 9 803 80
AK022988.1 - 3 - 4 285 80
AY596970.1 - 8 - 9 1194 80
AY596971.1 - 8 - 9 1194 80
BC016305.1 - 3 - 4 338 80
BC150596.1 - 8 - 9 902 80

View Full Product Details