Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC200830
- Entrez Gene ID
-
200830
- Gene Name
- uncharacterized LOC200830
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK054953.1 |
- |
1 - 1 |
1752 |
101 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB325910.1 |
- |
1 |
|
|
Chr. 3 - 161265627 - 161266154 [-] on Build GRCh38 |
101 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
NMD3 ribosome export adaptor |
NMD3 |
CGI-07 |
-11.6kb
|
Sense |
Chr. - 161221262 - 161253532 on Build GRCh38 |
Downstream |
serine palmitoyltransferase small subunit B |
SPTSSB |
ADMP, C3orf57, SSSPTB |
78.6kb
|
Anti-sense |
Chr. - 161344784 - 161373123 on Build GRCh38 |
View Full Product Details