Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SOCS7
- Entrez Gene ID
-
30837
- Gene Name
- suppressor of cytokine signaling 7
- Gene Aliases
-
NAP4, NCKAP4
- Chromosome Location
-
Chr.17: 38351876 - 38405593 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU629609.1 |
- |
1 |
|
|
Chr. 17 - 38402112 - 38402760 [-] on Build GRCh38 |
56 |
GenBankmRNA |
BE249857.1 |
- |
|
136 |
|
Chr. - - [-] on Build |
56 |
GenBankmRNA |
BI255871.1 |
- |
1 |
296 |
|
Chr. 17 - 38401911 - 38402783 [+] on Build GRCh38 |
56 |
GenBankmRNA |
BE293552.1 |
- |
1 |
137 |
|
Chr. 17 - 38402071 - 38402484 [+] on Build GRCh38 |
56 |
GenBankmRNA |
HY263203.1 |
- |
1 |
|
|
Chr. 17 - 38402054 - 38402532 [-] on Build GRCh38 |
56 |
GenBankmRNA |
BG329927.1 |
- |
|
240 |
|
Chr. - - [-] on Build |
56 |
GenBankmRNA |
DA834203.1 |
- |
1 |
469 |
|
Chr. 17 - 38401739 - 38402438 [+] on Build GRCh38 |
56 |
GenBankmRNA |
BE796289.1 |
- |
1 |
477 |
|
Chr. 17 - 38401731 - 38402507 [+] on Build GRCh38 |
56 |
GenBankmRNA |
BF691440.1 |
- |
1 |
351 |
|
Chr. 17 - 38401856 - 38402594 [+] on Build GRCh38 |
56 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
G protein-coupled receptor 179 |
GPR179 |
CSNB1E, GPR158L, GPR158L1 |
-56.4kb
|
Anti-sense |
Chr. - 38325610 - 38343810 on Build GRCh38 |
Downstream |
Rho GTPase activating protein 23 |
ARHGAP23 |
|
16.6kb
|
Sense |
Chr. - 38419286 - 38512392 on Build GRCh38 |
View Full Product Details