The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AY102071.1 | - | 3 - 4 | 303 | 79 |
BC129834.1 | - | 3 - 4 | 669 | 79 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | developmental pluripotency associated 3 pseudogene 2 | DPPA3P2 | STELLAR | -104.6kb | Anti-sense | Chr. - 36371318 - 36372369 on Build GRCh38 |
Downstream | NK2 homeobox 1 | NKX2-1 | BCH, BHC, NK-2, NKX2.1, NKX2A, NMTC1, T/EBP, TEBP, TITF1, TTF-1, TTF1 | 38.5kb | Sense | Chr. - 36516397 - 36520225 on Build GRCh38 |