Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PHLPP2
- Entrez Gene ID
-
23035
- Gene Name
- PH domain and leucine rich repeat protein phosphatase 2
- Gene Aliases
-
PHLPPL, PPM3B
- Chromosome Location
-
Chr.16: 71644924 - 71724701 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX648394.1 |
- |
2 - 2 |
1844 |
106 |
- Gene Symbol
- MARVELD3
- Entrez Gene ID
-
91862
- Gene Name
- MARVEL domain containing 3
- Gene Aliases
-
MARVD3, MRVLDC3
- Chromosome Location
-
Chr.16: 71626137 - 71641965 on Build GRCh38
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tyrosine aminotransferase |
TAT |
|
-60.7kb
|
Sense |
Chr. - 71566851 - 71577095 on Build GRCh38 |
View Full Product Details