Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TCTN1
- Entrez Gene ID
-
79600
- Gene Name
- tectonic family member 1
- Gene Aliases
-
JBTS13, TECT1
- Chromosome Location
-
Chr.12: 110614027 - 110649130 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC030993.1 |
- |
1 - 1 |
48 |
83 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG399158.1 |
- |
1 |
48 |
|
Chr. 12 - 110632450 - 110633076 [+] on Build GRCh38 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
PTC7 protein phosphatase homolog |
PPTC7 |
TA-PP2C, TAPP2C |
-49.2kb
|
Anti-sense |
Chr. - 110534432 - 110583259 on Build GRCh38 |
Downstream |
hydrogen voltage gated channel 1 |
HVCN1 |
HV1, VSOP |
15.0kb
|
Anti-sense |
Chr. - 110648686 - 110704945 on Build GRCh38 |
View Full Product Details