Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD20A11P
Entrez Gene ID
391267
Gene Name
ankyrin repeat domain 20 family member A11, pseudogene
Gene Aliases
C21orf81
Chromosome Location
Chr.21: 13943769 - 13980444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027270.1 5 - 6 1000 72
GenBank mRNA AF426257.1 - 5 - 6 1000 72


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI824886.1 - 1 - 2 Chr. 21 - 13943782 - 13979808 [+] on Build GRCh38 72
GenBankmRNA AI651100.1 - 1 - 2 Chr. 21 - 13943782 - 13979812 [+] on Build GRCh38 72
GenBankmRNA AI978581.1 - 1 - 2 Chr. 21 - 13943782 - 13979835 [+] on Build GRCh38 72
GenBankmRNA AI651101.1 - 1 - 2 Chr. 21 - 13943782 - 13979809 [+] on Build GRCh38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream cytochrome P450 family 4 subfamily F member 29, pseudogene CYP4F29P 4F-se4[6:7:8], C21orf15, CYP4F-se4[6:7:8], CYP4F3LP -95.4kb Sense Chr. - 13843133 - 13848364 on Build GRCh38
Downstream uncharacterized LOC105377134 LOC105377134 82.3kb Anti-sense Chr. - 14027399 - 14060948 on Build GRCh38

View Full Product Details