Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTF2IRD2
Entrez Gene ID
84163
Gene Name
GTF2I repeat domain containing 2
Gene Aliases
FP630, GTF2IRD2 alpha, GTF2IRD2A
Chromosome Location
Chr.7: 74796144 - 74851576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173537.3 NP_775808.3 13 - 14 1246 72
GenBank mRNA AK095257.1 - 10 - 11 805 72
AK225845.1 - 13 - 14 1203 72
AK291431.1 - 13 - 14 1215 72
AY260739.1 - 13 - 14 1199 72
AY312853.1 - 13 - 14 1199 72
AY312854.1 - 12 - 13 1039 72

Gene Symbol
GTF2IRD2B
Entrez Gene ID
389524
Gene Name
GTF2I repeat domain containing 2B
Gene Aliases
-
Chromosome Location
Chr.7: 75092556 - 75149817 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003795.2 NP_001003795.1 13 - 14 1246 72
GenBank mRNA AK095572.1 - 4 - 5 298 72
AY312850.1 - 13 - 14 1221 72
AY312851.1 - 12 - 13 1146 72
BC067859.1 - 13 - 14 1211 72
BC136849.1 - 13 - 14 1206 72
BX647624.1 - 7 - 8 719 72

Gene Symbol
GTF2IRD2P1
Entrez Gene ID
401375
Gene Name
GTF2I repeat domain containing 2 pseudogene 1
Gene Aliases
GTF2IRD2P
Chromosome Location
Chr.7: 73242751 - 73280119 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY312852.1 - 10 - 11 793 72

View Full Product Details