Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TATDN1
Entrez Gene ID
83940
Gene Name
TatD DNase domain containing 1
Gene Aliases
CDA11
Chromosome Location
Chr.8: 124488494 - 124539088 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146160.1 NP_001139632.1 9 - 10 711 98
NM_001317889.1 NP_001304818.1 12 - 13 935 98
NM_001317890.1 NP_001304819.1 9 - 10 810 98
NM_001317891.1 NP_001304820.1 8 - 9 574 98
NM_032026.3 NP_114415.1 11 - 12 827 98
NR_027427.1 12 - 13 939 98
XM_006716666.3 XP_006716729.1 12 - 13 955 98
XM_006716667.3 XP_006716730.1 12 - 13 1000 98
XM_006716668.3 XP_006716731.1 12 - 13 1013 98
XM_006716669.3 XP_006716732.1 10 - 11 897 98
XM_006716670.3 XP_006716733.1 11 - 12 856 98
XM_006716671.3 XP_006716734.1 10 - 11 826 98
XM_011517331.2 XP_011515633.1 11 - 12 851 98
XM_017013895.1 XP_016869384.1 11 - 12 837 98
XM_017013896.1 XP_016869385.1 11 - 12 892 98
XM_017013897.1 XP_016869386.1 10 - 11 813 98
XM_017013898.1 XP_016869387.1 10 - 11 750 98
GenBank mRNA AF212250.1 - 11 - 12 810 98
AK094861.1 - 3 - 4 1827 98
AK312204.1 - 11 - 12 823 98
AY071865.1 - 9 - 10 700 98
BC064964.1 - 11 - 12 811 98
BU784642.1 - 1 - 2 98
BX332564.2 - 12 - 13 918 98

View Full Product Details