Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
KCNE3 | NM_005472.4 | NP_005463.1 | ||
XM_011544713.2 | XP_011543015.1 | |||
XM_017017047.1 | XP_016872536.1 | |||
XM_017017048.1 | XP_016872537.1 | |||
XM_017017050.1 | 1 | 284 | XP_016872539.1 | |
XM_017017051.1 | XP_016872540.1 | |||
XM_017017052.1 | XP_016872541.1 | |||
CD709836.1 | ||||
CR936625.1 | CAI56768.1 | |||
DQ192291.1 | ABB00304.1 | |||
DR006135.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34979 | Chr11:74167643 - 74822407 on Build GRCh38 | Gain | LOC101928580 PGM2L1 MIR4696 POLD3 MIR548AL LOC100287896 C2CD3 KCNE3 P4HA3 LIPT2 RNF169 PPME1 CHRDL2 |
esv2759837 | Chr11:74131564 - 74846552 on Build GRCh38 | Gain | LOC101928580 PGM2L1 MIR4696 POLD3 MIR548AL LOC100287896 C2CD3 KCNE3 XRRA1 P4HA3 LIPT2 RNF169 PPME1 CHRDL2 |
nsv555443 | Chr11:74398121 - 74469444 on Build GRCh38 | Loss | PGM2L1 MIR548AL KCNE3 |
nsv832209 | Chr11:74444838 - 74618074 on Build GRCh38 | Gain | LIPT2 POLD3 LOC100287896 KCNE3 |
dgv209e214 | Chr11:74225600 - 74828943 on Build GRCh38 | Gain | P4HA3 LOC101928580 PGM2L1 MIR4696 LIPT2 POLD3 RNF169 MIR548AL LOC100287896 PPME1 CHRDL2 KCNE3 |
nsv393 | Chr11:74424817 - 74470372 on Build GRCh38 | Deletion | KCNE3 |
nsv825994 | Chr11:74225622 - 74823410 on Build GRCh38 | Gain | P4HA3 LOC101928580 PGM2L1 MIR4696 LIPT2 POLD3 RNF169 MIR548AL LOC100287896 PPME1 CHRDL2 KCNE3 |