Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC37A2
Entrez Gene ID
474170
Gene Name
leucine rich repeat containing 37 member A2
Gene Aliases
LRRC37
Chromosome Location
Chr.17: 46512657 - 46556747 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006607.2 NP_001006608.2 3 - 4 2750 97
XM_011524842.2 XP_011523144.1 3 - 4 2803 97
XM_011524843.2 XP_011523145.1 3 - 4 2803 97
XM_011524849.1 XP_011523151.1 3 - 4 2803 97
XM_011524850.2 XP_011523152.1 3 - 4 2803 97
GenBank mRNA AK126994.1 - 3 - 4 1432 97
AY386262.1 - 3 - 4 1171 97
BC144427.1 - 3 - 4 413 97
BC144428.1 - 3 - 4 413 97
JQ790563.1 - 3 - 4 307 97
JQ790564.1 - 3 - 4 307 97
JQ790568.1 - 3 - 4 307 97
JQ790571.1 - 3 - 4 307 97
JQ790576.1 - 3 - 4 307 97
JQ790584.1 - 3 - 4 307 97
JQ790606.1 - 3 - 4 307 97
JQ790610.1 - 3 - 4 307 97
JQ790617.1 - 3 - 4 307 97
JQ790626.1 - 3 - 4 307 97
JQ790628.1 - 3 - 4 307 97
JQ790700.1 - 3 - 4 307 97
JQ790703.1 - 3 - 4 283 97
JQ790735.1 - 3 - 4 309 97
JQ790739.1 - 3 - 4 309 97
JQ790740.1 - 3 - 4 309 97
JQ790750.1 - 3 - 4 309 97
JQ790764.1 - 3 - 4 309 97

Gene Symbol
LRRC37A
Entrez Gene ID
9884
Gene Name
leucine rich repeat containing 37A
Gene Aliases
LRRC37
Chromosome Location
Chr.17: 46248044 - 46339367 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014834.4 NP_055649.4 3 - 4 2750 97
XM_005257879.4 XP_005257936.1 3 - 4 3008 97
XM_005257880.4 XP_005257937.1 3 - 4 3008 97
XM_005257882.4 XP_005257939.1 4 - 5 3194 97
XM_005257883.4 XP_005257940.1 3 - 4 3008 97
XM_005257884.4 XP_005257941.1 3 - 4 3008 97
XM_017025472.1 XP_016880961.1 5 - 6 2607 97
XM_017025473.1 XP_016880962.1 3 - 4 254 97
GenBank mRNA BC040501.1 - 3 - 4 2750 97
CR749232.1 - 3 - 4 2751 97
JQ790562.1 - 3 - 4 307 97
JQ790566.1 - 3 - 4 307 97
JQ790572.1 - 3 - 4 307 97
JQ790577.1 - 3 - 4 307 97
JQ790578.1 - 3 - 4 307 97
JQ790579.1 - 3 - 4 307 97
JQ790580.1 - 3 - 4 307 97
JQ790581.1 - 3 - 4 307 97
JQ790688.1 - 3 - 4 307 97
JQ790690.1 - 3 - 4 307 97
JQ790691.1 - 3 - 4 307 97
JQ790692.1 - 3 - 4 307 97
JQ790693.1 - 3 - 4 307 97
JQ790694.1 - 3 - 4 307 97
JQ790699.1 - 3 - 4 307 97
JQ790726.1 - 3 - 4 309 97
JQ790727.1 - 3 - 4 309 97
JQ790729.1 - 3 - 4 309 97
JQ790730.1 - 3 - 4 309 97
JQ790738.1 - 3 - 4 309 97
JQ790741.1 - 3 - 4 309 97
JQ790743.1 - 3 - 4 309 97
JQ790744.1 - 3 - 4 308 97
JQ790749.1 - 3 - 4 309 97
JQ790752.1 - 3 - 4 309 97
JQ790755.1 - 3 - 4 309 97
JQ790756.1 - 3 - 4 309 97
JQ790757.1 - 3 - 4 309 97
JQ790758.1 - 3 - 4 309 97

Gene Symbol
LRRC37A3
Entrez Gene ID
374819
Gene Name
leucine rich repeat containing 37 member A3
Gene Aliases
LRRC37, LRRC37A
Chromosome Location
Chr.17: 64854147 - 64966228 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199340.3 NP_955372.2 6 - 7 3413 97
XM_011524776.1 XP_011523078.1 3 - 4 852 97
XM_011524778.2 XP_011523080.2 4 - 5 377 97
XM_017024593.1 XP_016880082.1 4 - 5 3242 97
XM_017024599.1 XP_016880088.1 5 - 6 2614 97
XM_017024600.1 XP_016880089.1 4 - 5 3242 97
XM_017024601.1 XP_016880090.1 3 - 4 554 97
XM_017024603.1 XP_016880092.1 3 - 4 852 97
XM_017024607.1 XP_016880096.1 3 - 4 157 97
XM_017024608.1 XP_016880097.1 3 - 4 1174 97
GenBank mRNA AB011135.2 - 5 - 6 3278 97
AK315945.1 - 3 - 4 885 97
BC043145.1 - 3 - 4 2755 97
BI560889.1 - 3 - 4 194 97
JQ790603.1 - 3 - 4 307 97
JQ790607.1 - 3 - 4 307 97
JQ790608.1 - 3 - 4 307 97
JQ790612.1 - 3 - 4 307 97
JQ790613.1 - 3 - 4 307 97
JQ790704.1 - 3 - 4 307 97
JQ790705.1 - 3 - 4 307 97
JQ790706.1 - 3 - 4 307 97
JQ790728.1 - 3 - 4 309 97
JQ790731.1 - 3 - 4 309 97
JQ790734.1 - 3 - 4 317 97
JQ790737.1 - 3 - 4 309 97
JQ790747.1 - 3 - 4 309 97
JQ790751.1 - 3 - 4 309 97
JQ790753.1 - 3 - 4 309 97
JQ790759.1 - 3 - 4 309 97
JQ790760.1 - 3 - 4 307 97
JQ790761.1 - 3 - 4 309 97
JQ790762.1 - 3 - 4 310 97
JQ790765.1 - 3 - 4 309 97
JQ790766.1 - 3 - 4 309 97

Gene Symbol
LRRC37A4P
Entrez Gene ID
55073
Gene Name
leucine rich repeat containing 37 member A4, pseudogene
Gene Aliases
LRRC37, LRRC37A4
Chromosome Location
Chr.17: 45505883 - 45520523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JQ790585.1 - 3 - 4 307 97
JQ790586.1 - 3 - 4 307 97
JQ790630.1 - 3 - 4 307 97
JQ790631.1 - 3 - 4 307 97
JQ790698.1 - 3 - 4 307 97

View Full Product Details