Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA8F
Entrez Gene ID
100132565
Gene Name
golgin A8 family member F
Gene Aliases
-
Chromosome Location
Chr.15: 28378638 - 28391266 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033351.1 4 - 5 431 102
GenBank mRNA AK057241.1 - 4 - 5 350 102
BC144672.1 - 2 - 3 128 102
BC150302.1 - 4 - 5 405 102
DB071191.1 - 4 - 5 431 102
DQ309038.1 - 4 - 5 320 102

Gene Symbol
GOLGA8CP
Entrez Gene ID
729786
Gene Name
golgin A8 family member C, pseudogene
Gene Aliases
GOLGA8C
Chromosome Location
Chr.15: 20562349 - 20575703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027411.2 4 - 5 431 102
GenBank mRNA DB085317.1 - 4 - 5 405 102

Gene Symbol
GOLGA8DP
Entrez Gene ID
100132979
Gene Name
golgin A8 family member D, pseudogene
Gene Aliases
GOLGA8D
Chromosome Location
Chr.15: 23157340 - 23170783 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027407.1 4 - 5 431 102
GenBank mRNA AK292534.1 - 4 - 5 317 102
BC110417.1 - 4 - 5 402 102

Gene Symbol
GOLGA8EP
Entrez Gene ID
390535
Gene Name
golgin A8 family member E, pseudogene
Gene Aliases
GOLGA8E
Chromosome Location
Chr.15: 22424673 - 22438026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033350.1 4 - 5 431 102
GenBank mRNA AK310240.1 - 4 - 5 405 102
BC063309.1 - 4 - 5 405 102
DQ309037.1 - 4 - 5 405 102

Gene Symbol
GOLGA8G
Entrez Gene ID
283768
Gene Name
golgin A8 family member G
Gene Aliases
-
Chromosome Location
Chr.15: 28519611 - 28532997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033353.1 4 - 5 431 102
GenBank mRNA AK302813.1 - 4 - 5 425 102
BC066774.1 - 4 - 5 405 102
BC125193.1 - 3 - 4 223 102
BC125194.1 - 3 - 4 223 102

Gene Symbol
GOLGA8S
Entrez Gene ID
653061
Gene Name
golgin A8 family member S
Gene Aliases
-
Chromosome Location
Chr.15: 23354748 - 23368324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_038843.1 4 - 5 405 102
GenBank mRNA DB053404.1 - 4 - 5 405 102

View Full Product Details