Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLEC4M
Entrez Gene ID
10332
Gene Name
C-type lectin domain family 4 member M
Gene Aliases
CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN
Chromosome Location
Chr.19: 7763052 - 7770559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144904.1 NP_001138376.1 4 - 5 751 78
NM_001144905.1 NP_001138377.1 5 - 6 832 78
NM_001144909.1 NP_001138381.1 5 - 6 766 78
NM_001144910.1 NP_001138382.1 5 - 6 835 78
NM_001144911.1 NP_001138383.1 3 - 4 820 78
NM_014257.4 NP_055072.3 4 - 5 904 78
XM_006722611.2 XP_006722674.1 4 - 5 836 78
XM_006722612.3 XP_006722675.1 3 - 4 910 78
XM_006722613.3 XP_006722676.1 3 - 4 912 78
XM_006722614.3 XP_006722677.1 2 - 3 828 78
XM_006722615.2 XP_006722678.1 4 - 5 904 78
GenBank mRNA AB015629.1 - 3 - 4 726 78
AF245219.2 - 4 - 5 810 78
AF290887.1 - 5 - 6 770 78
AK292278.1 - 5 - 6 831 78
AK304499.1 - 4 - 5 684 78
AK314704.1 - 5 - 6 765 78
AY042234.1 - 4 - 5 786 78
AY343913.1 - 5 - 6 649 78
BC038851.1 - 4 - 5 858 78
BC110614.2 - 2 - 3 759 78

View Full Product Details