Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KPNA7
Entrez Gene ID
402569
Gene Name
karyopherin subunit alpha 7
Gene Aliases
IPOA8
Chromosome Location
Chr.7: 99145467 - 99219417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011516215.2 XP_011514517.1 9 - 10 1672 92
XM_017012208.1 XP_016867697.1 9 - 10 1672 92
XM_017012209.1 XP_016867698.1 9 - 10 1660 92
XM_017012210.1 XP_016867699.1 9 - 10 1748 92
XM_017012211.1 XP_016867700.1 9 - 10 1672 92
XM_017012212.1 XP_016867701.1 9 - 10 1462 92
XM_017012213.1 XP_016867702.1 9 - 10 1800 92
XM_017012214.1 XP_016867703.1 9 - 10 1376 92
XM_017012216.1 XP_016867705.1 6 - 7 1120 92
GenBank mRNA EU126604.1 - 8 - 9 1316 92

View Full Product Details