Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCOC
Entrez Gene ID
60592
Gene Name
short coiled-coil protein
Gene Aliases
HRIHFB2072, SCOCO, UNC-69
Chromosome Location
Chr.4: 140257286 - 140382556 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001153635.1 NP_001147107.1 4 - 5 263 126
NM_001153663.1 NP_001147135.1 3 - 4 356 126
GenBank mRNA BC062684.1 - 4 - 5 263 126

View Full Product Details