Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCRL1
Entrez Gene ID
115350
Gene Name
Fc receptor like 1
Gene Aliases
CD307a, FCRH1, IFGP1, IRTA5
Chromosome Location
Chr.1: 157792249 - 157820150 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159397.1 NP_001152869.1 7 - 8 1112 74
XM_005244866.2 XP_005244923.1 8 - 9 1200 74
XM_011509128.1 XP_011507430.1 8 - 9 1200 74
XM_011509129.1 XP_011507431.1 8 - 9 1200 74
XM_011509130.1 XP_011507432.1 8 - 9 1200 74
GenBank mRNA AK096690.1 - 7 - 8 1051 74
AL833674.1 - 4 - 5 283 74

View Full Product Details