Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCRL1
Entrez Gene ID
115350
Gene Name
Fc receptor like 1
Gene Aliases
CD307a, FCRH1, IFGP1, IRTA5
Chromosome Location
Chr.1: 157792249 - 157820150 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159397.1 NP_001152869.1 9 - 10 1203 88
XM_005244866.2 XP_005244923.1 10 - 11 1291 88
XM_011509125.1 XP_011507427.1 11 - 12 1299 88
XM_011509126.1 XP_011507428.1 11 - 12 1296 85
XM_011509128.1 XP_011507430.1 10 - 11 1303 100
XM_011509129.1 XP_011507431.1 10 - 11 1300 97
XM_011509130.1 XP_011507432.1 10 - 11 1288 85
XM_011509131.1 XP_011507433.1 11 - 12 1311 100
XM_011509132.2 XP_011507434.1 11 - 12 1308 97
GenBank mRNA AK096690.1 - 9 - 10 1142 88
AL833674.1 - 6 - 7 374 88

View Full Product Details