Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
KRT83 | NM_002282.3 | NP_002273.3 | ||
AK314904.1 | ||||
BC069546.1 | AAH69546.1 | |||
BC126935.1 | ||||
BC128063.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1503n100 | Chr12:52285116 - 52394255 on Build GRCh38 | Gain | KRT81 KRT85 KRT83 KRT86 KRT84 KRT82 |
esv2759899 | Chr12:52169480 - 52391735 on Build GRCh38 | Gain | KRT7 KRT81 LINC00592 KRT85 KRT83 C12orf80 KRT86 KRT84 KRT80 KRT87P |
dgv29e180 | Chr12:52291976 - 52389509 on Build GRCh38 | Gain | KRT85 KRT83 KRT86 KRT84 |
nsv516938 | Chr12:52283925 - 52394228 on Build GRCh38 | Gain+Loss | KRT81 KRT85 KRT83 KRT86 KRT84 KRT82 |
dgv487e212 | Chr12:52294699 - 52388067 on Build GRCh38 | Gain | KRT85 KRT83 KRT86 KRT84 |
dgv200n27 | Chr12:52297319 - 52395144 on Build GRCh38 | Gain | KRT85 KRT83 KRT86 KRT84 KRT82 |
dgv9e209 | Chr12:52294216 - 52390216 on Build GRCh38 | Duplication | KRT85 KRT83 KRT86 KRT84 |
dgv2630n54 | Chr12:52287930 - 52395144 on Build GRCh38 | Gain | KRT81 KRT85 KRT83 KRT86 KRT84 KRT82 |
esv27319 | Chr12:52287366 - 52389709 on Build GRCh38 | Gain+Loss | KRT81 KRT85 KRT83 KRT86 KRT84 |
dgv70e55 | Chr12:52282619 - 52398149 on Build GRCh38 | Gain | KRT81 KRT85 KRT83 KRT86 KRT84 KRT82 |
esv2760832 | Chr12:52294900 - 52389052 on Build GRCh38 | Gain | KRT85 KRT83 KRT86 KRT84 |