Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LUNAR1
Entrez Gene ID
104564224
Gene Name
leukemia-associated non-coding IGF1R activator RNA 1
Gene Aliases
-
Chromosome Location
Chr.15: 99014704 - 99031050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL109706.1 - 1 - 1 2243 85


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB346252.1 - 1 Chr. 15 - 99030492 - 99031044 [-] on Build GRCh38 85
GenBankmRNA AA001199.1 - 1 Chr. 15 - 99030441 - 99031044 [-] on Build GRCh38 85
GenBankmRNA AA001380.1 - 1 Chr. 15 - 99030522 - 99031044 [-] on Build GRCh38 85


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream pyroglutamyl-peptidase I-like PGPEP1L -20.7kb Anti-sense Chr. - 98968230 - 99007795 on Build GRCh38
Downstream synemin SYNM DMN, SYN 74.0kb Sense Chr. - 99105069 - 99136586 on Build GRCh38

View Full Product Details