Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FAM84A
- Entrez Gene ID
-
151354
- Gene Name
- family with sequence similarity 84 member A
- Gene Aliases
-
NSE1, PP11517
- Chromosome Location
-
Chr.2: 14632686 - 14640046 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA778134.1 |
- |
1 |
|
|
Chr. 2 - 14639656 - 14640043 [-] on Build GRCh38 |
102 |
GenBankmRNA |
H89896.1 |
- |
1 |
|
|
Chr. 2 - 14639781 - 14640046 [+] on Build GRCh38 |
103 |
GenBankmRNA |
AI264453.1 |
- |
1 |
|
|
Chr. 2 - 14639672 - 14640042 [-] on Build GRCh38 |
102 |
GenBankmRNA |
AA922936.1 |
- |
1 |
|
|
Chr. 2 - 14639572 - 14640042 [-] on Build GRCh38 |
102 |
GenBankmRNA |
AI264428.1 |
- |
1 |
|
|
Chr. 2 - 14639734 - 14640042 [-] on Build GRCh38 |
102 |
GenBankmRNA |
BQ027984.1 |
- |
1 |
|
|
Chr. 2 - 14639629 - 14640046 [-] on Build GRCh38 |
102 |
GenBankmRNA |
W74462.1 |
- |
|
|
|
Chr. - - [-] on Build |
102 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 276 |
LINC00276 |
NCRNA00276 |
-238.8kb
|
Anti-sense |
Chr. - 14228874 - 14400958 on Build GRCh38 |
Downstream |
neuroblastoma amplified sequence |
NBAS |
ILFS2, NAG, SOPH |
526.9kb
|
Anti-sense |
Chr. - 14998067 - 15561348 on Build GRCh38 |
View Full Product Details