Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA G36890.1 - 1 43 Chr. 6 - 45334104 - 45334452 [-] on Build GRCh38 90
GenBankmRNA N64274.1 - 1 43 Chr. 6 - 45334103 - 45334452 [-] on Build GRCh38 90


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream microRNA 586 MIR586 MIRN586, hsa-mir-586 -136.3kb Sense Chr. - 45197674 - 45197770 on Build GRCh38
Downstream uncharacterized LOC105375077 LOC105375077 220.7kb Sense Chr. - 45554330 - 45576791 on Build GRCh38

View Full Product Details