Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AL047449.1 |
- |
1 |
|
|
Chr. 1 - 178480771 - 178481209 [-] on Build GRCh38 |
84 |
GenBankmRNA |
AF075003.1 |
- |
1 |
451 |
|
Chr. 1 - 178480607 - 178481205 [+] on Build GRCh38 |
84 |
GenBankmRNA |
R37813.1 |
- |
|
|
|
Chr. - - [-] on Build |
84 |
GenBankmRNA |
BX094279.1 |
- |
1 |
449 |
|
Chr. 1 - 178480608 - 178481209 [+] on Build GRCh38 |
84 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 83 |
LINC00083 |
NCRNA00083 |
|
Anti-sense |
Chr. - 178479514 - 178499623 on Build GRCh38 |
Downstream |
testis expressed 35 |
TEX35 |
C1orf49, TSC24 |
31.9kb
|
Sense |
Chr. - 178513077 - 178523500 on Build GRCh38 |
View Full Product Details