Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA H52528.1 - 1 Chr. 17 - 40925645 - 40925949 [+] on Build GRCh38 72
GenBankmRNA AF075083.1 - 1 344 Chr. 17 - 40925606 - 40926163 [-] on Build GRCh38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream keratin 20 KRT20 CD20, CK-20, CK20, K20, KRT21 -40.4kb Sense Chr. - 40875889 - 40885243 on Build GRCh38
Downstream keratin 39 KRT39 CK-39, K39, KA35 32.3kb Sense Chr. - 40958417 - 40966892 on Build GRCh38

View Full Product Details