The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_110818.1 | 2 - 3 | 1545 | 75 | |
GenBank mRNA | BC069263.1 | - | 2 - 3 | 1545 | 75 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 92B | CCDC92B | -212.7kb | Sense | Chr. - 2720801 - 2748500 on Build GRCh38 | |
Downstream | olfactory receptor family 1 subfamily D member 5 | OR1D5 | C17orf2, OR17-2, OR17-30, OR17-31, OR1D4 | 98.7kb | Sense | Chr. - 3062669 - 3063607 on Build GRCh38 |