Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100293044
Entrez Gene ID
100293044
Gene Name
uncharacterized LOC100293044
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK297180.1 - 1 - 1 462 66


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DC404395.1 - 1 461 Chr. 19 - 34676988 - 34677581 [-] on Build GRCh38 66
GenBankmRNA DC349842.1 - 1 462 Chr. 19 - 34677077 - 34677581 [-] on Build GRCh38 66
GenBankmRNA DC330563.1 - 1 169 Chr. 19 - 34676724 - 34677290 [-] on Build GRCh38 66
GenBankmRNA DC368284.1 - 1 141 Chr. 19 - 34676725 - 34677259 [-] on Build GRCh38 67
GenBankmRNA DC295468.1 - 1 181 Chr. 19 - 34676840 - 34677299 [-] on Build GRCh38 66
GenBankmRNA BX490537.1 - 1 151 Chr. 19 - 34676700 - 34677262 [-] on Build GRCh38 66
GenBankmRNA DC319368.1 - 1 183 Chr. 19 - 34676717 - 34677301 [-] on Build GRCh38 66
GenBankmRNA DC325815.1 - 1 123 Chr. 19 - 34676663 - 34677241 [-] on Build GRCh38 67
GenBankmRNA CR603799.1 - 1 150 Chr. 19 - 34675717 - 34677267 [-] on Build GRCh38 66
GenBankmRNA DC328351.1 - 1 117 Chr. 19 - 34676658 - 34677235 [-] on Build GRCh38 66
GenBankmRNA DC302608.1 - 1 135 Chr. 19 - 34676678 - 34677251 [-] on Build GRCh38 67
GenBankmRNA BX342528.2 - 1 149 Chr. 19 - 34676292 - 34677267 [-] on Build GRCh38 66
GenBankmRNA DC351433.1 - 1 138 Chr. 19 - 34676692 - 34677259 [-] on Build GRCh38 66
GenBankmRNA DC332278.1 - 1 132 Chr. 19 - 34676689 - 34677251 [-] on Build GRCh38 66
GenBankmRNA AA833672.1 - 1 Chr. 19 - 34676902 - 34677244 [+] on Build GRCh38 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream secretoglobin family 2B member 3, pseudogene SCGB2B3P C2B, SCGB4A3P -29.7kb Anti-sense Chr. - 34645873 - 34646052 on Build GRCh38
Downstream zinc finger protein 302 ZNF302 HSD16, MST154, MSTP154, ZNF135L, ZNF140L, ZNF327 372.0 Anti-sense Chr. - 34677619 - 34686397 on Build GRCh38

View Full Product Details