Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY225932.1 - 1 Chr. 11 - 94868527 - 94868944 [-] on Build GRCh38 68
GenBankmRNA AK093646.1 - 1 1753 Chr. 11 - 94867068 - 94868944 [+] on Build GRCh38 68
GenBankmRNA BX110149.1 - 1 Chr. 11 - 94868490 - 94868958 [-] on Build GRCh38 68
GenBankmRNA DB342809.1 - 1 Chr. 11 - 94868356 - 94868944 [-] on Build GRCh38 68


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC105369438 LOC105369438 -126.7kb Anti-sense Chr. - 94545332 - 94740355 on Build GRCh38
Downstream CWC15 spliceosome-associated protein CWC15 AD002, C11orf5, Cwf15, HSPC148, ORF5 93.7kb Anti-sense Chr. - 94962622 - 94973612 on Build GRCh38

View Full Product Details