Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW937711.1 |
- |
1 |
|
|
Chr. 19 - 11894682 - 11895279 [-] on Build GRCh38 |
97 |
GenBankmRNA |
AK093694.1 |
- |
1 |
948 |
|
Chr. 19 - 11893956 - 11896217 [+] on Build GRCh38 |
97 |
GenBankmRNA |
H71945.1 |
- |
1 |
|
|
Chr. 19 - 11894701 - 11895099 [+] on Build GRCh38 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger protein 439 |
ZNF439 |
|
-24.5kb
|
Sense |
Chr. - 11866029 - 11869491 on Build GRCh38 |
Downstream |
zinc finger protein 700 |
ZNF700 |
|
28.9kb
|
Sense |
Chr. - 11925068 - 11950773 on Build GRCh38 |
View Full Product Details