Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DENND1B
- Entrez Gene ID
-
163486
- Gene Name
- DENN domain containing 1B
- Gene Aliases
-
C1ORF18, C1orf218, FAM31B
- Chromosome Location
-
Chr.1: 197504748 - 197782138 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB244207.1 |
- |
1 |
234 |
|
Chr. 1 - 197506544 - 197507107 [-] on Build GRCh38 |
90 |
GenBankmRNA |
DB249531.1 |
- |
1 |
144 |
|
Chr. 1 - 197506456 - 197507018 [-] on Build GRCh38 |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
crumbs 1, cell polarity complex component |
CRB1 |
LCA8, RP12 |
-26.3kb
|
Anti-sense |
Chr. - 197201462 - 197478455 on Build GRCh38 |
Downstream |
chromosome 1 open reading frame 53 |
C1orf53 |
|
395.5kb
|
Anti-sense |
Chr. - 197902552 - 197907367 on Build GRCh38 |
View Full Product Details