Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MITF
- Entrez Gene ID
-
4286
- Gene Name
- melanogenesis associated transcription factor
- Gene Aliases
-
CMM8, MI, WS2, WS2A, bHLHe32
- Chromosome Location
-
Chr.3: 69739435 - 69968337 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY632572.1 |
- |
1 - 1 |
59 |
94 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
HY173803.1 |
- |
1 |
53 |
|
Chr. 3 - 69838514 - 69937934 [+] on Build GRCh38 |
94 |
GenBankmRNA |
HY180835.1 |
- |
1 |
53 |
|
Chr. 3 - 69838512 - 69937953 [+] on Build GRCh38 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
FERM domain containing 4B |
FRMD4B |
6030440G05Rik, GRSP1 |
-297.2kb
|
Anti-sense |
Chr. - 69168782 - 69541340 on Build GRCh38 |
Downstream |
survival associated mitochondrial melanoma specific oncogenic non-coding RNA |
SAMMSON |
LINC01212 |
161.1kb
|
Sense |
Chr. - 69999744 - 70015301 on Build GRCh38 |
View Full Product Details