The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022358.3 | NP_071753.2 | 2 - 2 | 1462 | 71 |
GenBank mRNA | BC082981.1 | - | 1 - 1 | 250 | 71 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | regulating synaptic membrane exocytosis 4 | RIMS4 | C20orf190, RIM 4, RIM-4, RIM4, RIM4-gamma, RIM4gamma | Anti-sense | Chr. - 44751804 - 44810338 on Build GRCh38 | |
Downstream | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta | YWHAB | GW128, HEL-S-1, HS1, KCIP-1, YWHAA | 133.3kb | Sense | Chr. - 44885599 - 44908532 on Build GRCh38 |