Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX384338.2 - 1 52 Chr. 4 - 52713378 - 52714137 [+] on Build GRCh38 118
GenBankmRNA CR611300.1 - 1 52 Chr. 4 - 52713378 - 52714137 [+] on Build GRCh38 118
GenBankmRNA BM685854.1 - 1 320 Chr. 4 - 52713116 - 52713681 [+] on Build GRCh38 118
GenBankmRNA CN411930.1 - 1 611 Chr. 4 - 52712823 - 52713547 [+] on Build GRCh38 118


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small nucleolar RNA, H/ACA box 26 SNORA26 HBI-6 -8.0 Sense Chr. - 52713249 - 52713370 on Build GRCh38
Downstream endogenous retrovirus group MER34 member 1 ERVMER34-1 24.8kb Anti-sense Chr. - 52743517 - 52751640 on Build GRCh38

View Full Product Details