Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AB075499.1 - 1 1344 Chr. 14 - 55554422 - 55556406 [-] on Build GRCh38 68


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream TATA-box binding protein like 2 TBPL2 TBP2, TRF3 -113.9kb Sense Chr. - 55414210 - 55440545 on Build GRCh38
Downstream KTN1 antisense RNA 1 KTN1-AS1 C14orf33, MYCLo-3 19.8kb Sense Chr. - 55576157 - 55580092 on Build GRCh38

View Full Product Details