Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK096407.1 - 1 1253 Chr. 9 - 134298463 - 134300413 [+] on Build GRCh38 68
GenBankmRNA AA586382.1 - 1 Chr. 9 - 134299519 - 134299897 [-] on Build GRCh38 68


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream RNA, U6atac small nuclear (U12-dependent splicing) RNU6ATAC RNU6ATAC1, U6ATAC -133.9kb Anti-sense Chr. - 134164440 - 134164564 on Build GRCh38
Downstream retinoid X receptor alpha RXRA NR2B1 26.1kb Sense Chr. - 134326463 - 134440586 on Build GRCh38

View Full Product Details