Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK098147.1 |
- |
2 - 3 |
1898 |
|
Chr. 22 - 30489075 - 30492804 [+] on Build GRCh38 |
104 |
GenBankmRNA |
DB348381.1 |
- |
1 - 2 |
|
|
Chr. 22 - 30490802 - 30492804 [-] on Build GRCh38 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
syndecan 4 pseudogene |
SDC4P |
|
-10.0kb
|
Anti-sense |
Chr. - 30481290 - 30481756 on Build GRCh38 |
Downstream |
SEC14 like lipid binding 6 |
SEC14L6 |
|
31.8kb
|
Anti-sense |
Chr. - 30522797 - 30546717 on Build GRCh38 |
View Full Product Details