Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIK3C3
Entrez Gene ID
5289
Gene Name
phosphatidylinositol 3-kinase catalytic subunit type 3
Gene Aliases
VPS34, Vps34, hVps34
Chromosome Location
Chr.18: 41955196 - 42084606 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC040982.1 - 1 - 1 2246 98


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA EB360799.1 - 1 462 Chr. 18 - 42083597 - 42084227 [+] on Build GRCh38 98
GenBankmRNA AW967911.1 - 1 Chr. 18 - 42083936 - 42084573 [-] on Build GRCh38 98
GenBankmRNA CA773164.1 - 1 315 Chr. 18 - 42083754 - 42084354 [+] on Build GRCh38 98
GenBankmRNA DA098595.1 - 1 311 Chr. 18 - 42083749 - 42084232 [+] on Build GRCh38 98


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream keratoconus gene 6 KC6 -561.2kb Anti-sense Chr. - 41480271 - 41520597 on Build GRCh38
Downstream long intergenic non-protein coding RNA 907 LINC00907 102.1kb Sense Chr. - 42186668 - 42691426 on Build GRCh38

View Full Product Details