Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC041910.1 - 1 843 Chr. 7 - 19416134 - 19418417 [+] on Build GRCh38 71


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream Fer3-like bHLH transcription factor FERD3L N-TWIST, NATO3, NTWIST, bHLHa31 -270.7kb Anti-sense Chr. - 19144782 - 19145421 on Build GRCh38
Downstream TWIST neighbor TWISTNB 277.0kb Anti-sense Chr. - 19695462 - 19709037 on Build GRCh38

View Full Product Details