Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK130115.1 - 1 848 Chr. 11 - 11513808 - 11514755 [-] on Build GRCh38 82


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream polypeptide N-acetylgalactosaminyltransferase 18 GALNT18 GALNACT18, GALNT15, GALNTL4, GalNAc-T15, GalNAc-T18 Sense Chr. - 11270874 - 11622045 on Build GRCh38
Downstream microRNA 4299 MIR4299 141.9kb Sense Chr. - 11656651 - 11656722 on Build GRCh38

View Full Product Details