Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PDK3
- Entrez Gene ID
-
5165
- Gene Name
- pyruvate dehydrogenase kinase 3
- Gene Aliases
-
CMTX6, GS1-358P8.4
- Chromosome Location
-
Chr.X: 24465227 - 24550466 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BF977927.1 |
- |
1 |
159 |
|
Chr. X - 24549682 - 24550314 [+] on Build GRCh38 |
87 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SPT20 homolog, SAGA complex component-like 1 |
SUPT20HL1 |
FAM48B1, SPT20L |
-180.5kb
|
Sense |
Chr. - 24362761 - 24365424 on Build GRCh38 |
Downstream |
phosphate cytidylyltransferase 1, choline, beta |
PCYT1B |
CCTB, CTB |
7.6kb
|
Anti-sense |
Chr. - 24558087 - 24672862 on Build GRCh38 |
View Full Product Details