Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ027602.1 - 1 Chr. 2 - 24826199 - 24826608 [-] on Build GRCh38 77
GenBankmRNA DB339766.1 - 1 Chr. 2 - 24821272 - 24826607 [-] on Build GRCh38 77
GenBankmRNA AI872373.1 - 1 Chr. 2 - 24826262 - 24826607 [-] on Build GRCh38 77
GenBankmRNA HY200275.1 - 1 Chr. 2 - 24826103 - 24826607 [-] on Build GRCh38 77
GenBankmRNA BC047034.1 - 1 846 Chr. 2 - 24825610 - 24826717 [+] on Build GRCh38 77


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream centromere protein O CENPO CENP-O, ICEN-36, MCM21R -3.2kb Sense Chr. - 24793115 - 24822376 on Build GRCh38
Downstream DnaJ heat shock protein family (Hsp40) member C27 DNAJC27 RBJ, RabJS 116.9kb Anti-sense Chr. - 24943636 - 24972094 on Build GRCh38

View Full Product Details