Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX647374.1 - 1 4154 Chr. 3 - 157483774 - 157488951 [-] on Build GRCh38 97


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ventricular zone expressed PH domain containing 1 VEPH1 MELT, VEPH Sense Chr. - 157259742 - 157503626 on Build GRCh38
Downstream PQ loop repeat containing 2 like PQLC2L C3orf55 54.4kb Anti-sense Chr. - 157543344 - 157601233 on Build GRCh38

View Full Product Details