Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TTBK2
- Entrez Gene ID
-
146057
- Gene Name
- tau tubulin kinase 2
- Gene Aliases
-
SCA11, TTBK
- Chromosome Location
-
Chr.15: 42738974 - 42920995 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB137885.1 |
- |
1 |
388 |
|
Chr. 15 - 42743017 - 42743551 [-] on Build GRCh38 |
92 |
GenBankmRNA |
DA808409.1 |
- |
1 |
117 |
|
Chr. 15 - 42742743 - 42743285 [-] on Build GRCh38 |
92 |
GenBankmRNA |
CN291889.1 |
- |
1 |
426 |
|
Chr. 15 - 42742993 - 42743594 [-] on Build GRCh38 |
92 |
GenBankmRNA |
DA498345.1 |
- |
1 |
316 |
|
Chr. 15 - 42742905 - 42743483 [-] on Build GRCh38 |
92 |
GenBankmRNA |
DA083763.1 |
- |
1 |
383 |
|
Chr. 15 - 42743105 - 42743550 [-] on Build GRCh38 |
92 |
GenBankmRNA |
CN356888.1 |
- |
|
102 |
|
Chr. - - [-] on Build |
92 |
GenBankmRNA |
DA057440.1 |
- |
1 |
76 |
|
Chr. 15 - 42742741 - 42743244 [-] on Build GRCh38 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
codanin 1 |
CDAN1 |
CDA1, CDAI, CDAN1A, DLT, PRO1295 |
-3.3kb
|
Sense |
Chr. - 42723557 - 42737219 on Build GRCh38 |
Downstream |
ubiquitin protein ligase E3 component n-recognin 1 |
UBR1 |
JBS |
198.6kb
|
Sense |
Chr. - 42942897 - 43106088 on Build GRCh38 |
View Full Product Details